Learn More
Bioworld Murashige Cattleya Orchid Multiplication Medium, 25 L
Catalog No. 50254963
Encompass_Preferred
Murashige Cattleya Orchid Multiplication MediumContains Macronutrients and Micronutrients and vitamins as described by Murashige & Skoog (1962), with sucrose 2%, agar 0.8%. Add 0.3 mg/L IAA, 1.75mg/L NAA and 1.75 mg/L BA (provided) after autoclaving.Provided as 1 unit to make 25L or 50L of medium, or 10 units to make 1L medium with 1 unit.Plant tissue culture testedWorking Concentration: 32.4 g/LpH: 2.75–3.75Soluble in water
Catalog No. 50-254-963 Supplier Bioworld Supplier No. 306301872
Murashige Cattleya Orchid Multiplication MediumContains Macronutrients and Micronutrients and vitamins as described by Murashige & Skoog (1962), with sucrose 2%, agar 0.8%. Add 0.3 mg/L IAA, 1.75mg/L NAA and 1.75 mg/L BA (provided) after autoclaving.Provided as 1 unit to make 25L or 50L of medium, or 10 units to make 1L medium with 1 unit.Plant tissue culture testedWorking Concentration: 32.4 g/LpH: 2.75–3.75Soluble in water