Learn More
Bioworld Murashige Fern Multiplication Medium, 50 L
Catalog No. 50254862
Encompass_Preferred
Murashige Fern Multiplication MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Linsmaier and Skoog (1965) with the addition of Sodium Phosphate Monobasic, Kinetin, and NAAContains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTASucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.66 g/LpH: 4.25–5.25Soluble in water
Catalog No. 50-254-862 Supplier Bioworld Supplier No. 306301494
Murashige Fern Multiplication MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Linsmaier and Skoog (1965) with the addition of Sodium Phosphate Monobasic, Kinetin, and NAAContains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTASucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.66 g/LpH: 4.25–5.25Soluble in water