Learn More
Bioworld Murashige Gerbera Multiplication Medium, 25 L
Catalog No. 50254864
Encompass_Preferred
Murashige Gerbera Multiplication MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) with the addition of Sodium Phosphate Monobasic, Adenine Hemisulfate, Kinetin, IAA, and L-TyrosineContains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTASucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.72 g/LSoluble in water
Catalog No. 50-254-864 Supplier Bioworld Supplier No. 306301503
Murashige Gerbera Multiplication MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) with the addition of Sodium Phosphate Monobasic, Adenine Hemisulfate, Kinetin, IAA, and L-TyrosineContains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTASucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.72 g/LSoluble in water