Learn More
Bioworld Murashige Kalanchoe Multiplication Medium, 25 L
Catalog No. 50254871
Encompass_Preferred
Murashige Kalanchoe Multiplication MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Linsmaier and Skoog (1965) with the addition of 2iP Contains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTASucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.41 g/LSoluble in water
Catalog No. 50-254-871 Supplier Bioworld Supplier No. 306301524
Add to Cart
Add to Cart
Murashige Kalanchoe Multiplication MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Linsmaier and Skoog (1965) with the addition of 2iP Contains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTASucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.41 g/LSoluble in water