Learn More
Bioworld Murashige Kalanchoe Pretransplant Medium, 25 L
Catalog No. 50254873
Encompass_Preferred
Murashige Kalanchoe Pretransplant MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Linsmaier and Skoog (1965) with the addition of IAA Contains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTASucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.41 g/LSoluble in water
Catalog No. 50-254-873 Supplier Bioworld Supplier No. 306301532
Murashige Kalanchoe Pretransplant MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Linsmaier and Skoog (1965) with the addition of IAA Contains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTASucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.41 g/LSoluble in water