Learn More
Bioworld Murashige & Miller Medium, 50 L
Catalog No. 50254735
Encompass_Preferred
Murashige & Miller MediumContains Micronutrients, Macronutrients, and Vitamins as described by Murashige andSkoog (1962)Syngonium Stage I and II Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-735 Supplier Bioworld Supplier No. 306300834
Murashige & Miller MediumContains Micronutrients, Macronutrients, and Vitamins as described by Murashige andSkoog (1962)Syngonium Stage I and II Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested