Learn More
Bioworld Murashige Modified Multiplication Medium, w/ Kinetin, 25 L
Catalog No. 50254876
Encompass_Preferred
Murashige Modified Multiplication Medium, KinetinContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Linsmaier and Skoog (1965) modified with the addition of IAA and KinetinContains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTAProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.41 g/LSoluble in water
Catalog No. 50-254-876 Supplier Bioworld Supplier No. 306301543
Murashige Modified Multiplication Medium, KinetinContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Linsmaier and Skoog (1965) modified with the addition of IAA and KinetinContains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTAProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.41 g/LSoluble in water