Learn More
PLANT CELL TECHNOLOGY Murashige & Skoog Basal Salts with Macronutrients, Micronutrients, Vitamins and Glycine, Plant Tissue Culture, MS Basal Salts Mod. 9, White to Yellow Powder, 50L Pouch
Catalog No. NC3905178
Encompass_Preferred
MS Basal Salts Mod. 9 is a plant tissue culture medium providing essential macro- and micronutrients for optimal plant growth. Supplied in a 50L single-use pouch, this off-white to yellow powdered media formulation supports tissue culture applications. The medium is fully soluble in water and forms a ready to autoclave solution, ensuring high reproducibility in research and commercial propagation. Ideal for laboratories, greenhouses, and educational applications. Stable under recommended storage conditions and suitable for integration into custom media preparations.
Catalog No. NC3905178 Supplier PLANT CELL TECHNOLOGY Supplier No. MSP0950LT.1
MS Basal Salts Mod. 9 is a plant tissue culture medium providing essential macro- and micronutrients for optimal plant growth. Supplied in a 50L single-use pouch, this off-white to yellow powdered media formulation supports tissue culture applications. The medium is fully soluble in water and forms a ready to autoclave solution, ensuring high reproducibility in research and commercial propagation. Ideal for laboratories, greenhouses, and educational applications. Stable under recommended storage conditions and suitable for integration into custom media preparations.