Learn More
Bioworld Murashige & Skoog/Gamborg (MG) Medium, 25 L
Catalog No. 50255016
Encompass_Preferred
MS Modified MediumContains the Macronutrients as described by Murashige and Skoog (1962) and Micronutrients as described by Gamborg et. al (1968) along with 0.83mg/L Potassium Iodide.Cytokines, Sucrose, and Auxins can be added as needed Sold in individual containers with enough medium to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 1.88 g/LSoluble in water
Catalog No. 50-255-016 Supplier Bioworld Supplier No. 306302062
MS Modified MediumContains the Macronutrients as described by Murashige and Skoog (1962) and Micronutrients as described by Gamborg et. al (1968) along with 0.83mg/L Potassium Iodide.Cytokines, Sucrose, and Auxins can be added as needed Sold in individual containers with enough medium to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 1.88 g/LSoluble in water