Learn More
Research Products International Corp Murashige & Skoog Medium with MES Buffer and Vitamins, 122.5 Grams of Powder, Makes 25 Liters of Solution
SDP
Catalog No. 50213510
Encompass_Preferred
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1969). MES buffer 50 mg/L added to prevent acidification during cultivation. Supplied as powder.
  • Concentration: 4905.19 mg/L
  • Hygroscopic
Catalog No. 50-213-510 Supplier Research Products International Corp Supplier No. M7080025.0
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1969). MES buffer 50 mg/L added to prevent acidification during cultivation. Supplied as powder.
  • Concentration: 4905.19 mg/L
  • Hygroscopic