Learn More
Research Products International Corp Murashige & Skoog Medium with MES Buffer and Vitamins, 24.5 Grams of Powder, Makes 5 Liters of Solution
SDP
Catalog No. 50213511
Encompass_Preferred
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1969). MES buffer 50 mg/L added to prevent acidification during cultivation. Supplied as powder.
  • Concentration: 4905.19 mg/L
  • Hygroscopic
Catalog No. 50-213-511 Supplier Research Products International Corp Supplier No. M708005.0
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1969). MES buffer 50 mg/L added to prevent acidification during cultivation. Supplied as powder.
  • Concentration: 4905.19 mg/L
  • Hygroscopic