Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Murashige & Skoog Shoot Multiplication Medium B, 4.5 Grams of Powder, Makes 1 Liter of Solution
SDP

Catalog No. 50213503
Encompass_Preferred

Contains micro and macro elements with vitamins as described by Huan & Murashige (1976) TCA manual.

  • Concentration: 4530.89 mg/L
  • Hygroscopic

Catalog No. 50-213-503 Supplier Research Products International Corp Supplier No. M705001.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains micro and macro elements with vitamins as described by Huan & Murashige (1976) TCA manual.

  • Concentration: 4530.89 mg/L
  • Hygroscopic

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.