Learn More
Bioworld Murashige and Skoog Medium, Micro Salt only, 50 L
Catalog No. 50255006
Encompass_Preferred
Murashige & Skoog (MS) Basal Salt MediumContains only micronutrients as described by Murashige & Skoog (1962)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 0.10 g/LpH: 3.8–4.8Soluble in water
Catalog No. 50-255-006 Supplier Bioworld Supplier No. 306302023
Murashige & Skoog (MS) Basal Salt MediumContains only micronutrients as described by Murashige & Skoog (1962)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 0.10 g/LpH: 3.8–4.8Soluble in water