Learn More
Alkali Scientific Murashige & Skoog Medium with MES Buffer, 24 Grams of Powder, Makes 5 Liters of Solution
SDP
Catalog No. 5000661612
Encompass_Preferred
Murashige & Skoog Medium with MES Buffer is designed for plant tissue culture and is available in a 5-liter size. This powder, containing 24 grams, makes 5 liters of solution, providing essential nutrients and pH stability with the inclusion of MES buffer. The buffer ensures the pH remains constant, which is vital for maintaining optimal growth conditions for plant cells. This medium is widely used in laboratories for plant cell culture, genetic studies, and regeneration research. MES buffer is particularly important in experiments that require precise pH control, making this medium an essential tool for researchers working with plant tissue cultures.
Catalog No. 50-006-61612 Supplier Alkali Scientific Supplier No. SHI005CP
Murashige & Skoog Medium with MES Buffer is designed for plant tissue culture and is available in a 5-liter size. This powder, containing 24 grams, makes 5 liters of solution, providing essential nutrients and pH stability with the inclusion of MES buffer. The buffer ensures the pH remains constant, which is vital for maintaining optimal growth conditions for plant cells. This medium is widely used in laboratories for plant cell culture, genetic studies, and regeneration research. MES buffer is particularly important in experiments that require precise pH control, making this medium an essential tool for researchers working with plant tissue cultures.