Learn More
Alkali Scientific Murashige and Skoog Medium with Vitamins, Van der Salm Modification, 4.4 Grams of Powder, Makes 1 Liter of Solution
SDP
Catalog No. 5000659713
Encompass_Preferred
Murashige & Skoog Medium with Vitamins, Van der Salm Modification, is a plant tissue culture medium designed for plant growth and regeneration studies. This 1-liter solution, made from 4.4 grams of powder, combines Murashige & Skoog’s base medium with essential vitamins and the Van der Salm modification. The modification optimizes the medium for plant species that require different nutrient levels for best growth. This medium is ideal for plant tissue culture, genetic studies, and plant regeneration, ensuring that plant cells receive the appropriate nutrients for development. Researchers often use this medium in experiments requiring a precise formulation to promote cell division and growth in plant tissues.
Catalog No. 50-006-59713 Supplier Alkali Scientific Supplier No. KOO001CP
Murashige & Skoog Medium with Vitamins, Van der Salm Modification, is a plant tissue culture medium designed for plant growth and regeneration studies. This 1-liter solution, made from 4.4 grams of powder, combines Murashige & Skoog’s base medium with essential vitamins and the Van der Salm modification. The modification optimizes the medium for plant species that require different nutrient levels for best growth. This medium is ideal for plant tissue culture, genetic studies, and plant regeneration, ensuring that plant cells receive the appropriate nutrients for development. Researchers often use this medium in experiments requiring a precise formulation to promote cell division and growth in plant tissues.