Learn More
Alkali Scientific Murashige and Skoog Medium with Vitamins, Van der Salm Modification, 44.6 Grams of Powder, Makes 10 Liters of Solution
SDP
Catalog No. 5000659866
Encompass_Preferred
Murashige & Skoog Medium with Vitamins, Van der Salm Modification, is a modified growth medium designed for large-scale plant tissue culture applications. With 44.6 grams of powder, it makes 10 liters of solution, supporting the growth and development of plant cells. The Van der Salm modification improves the medium for specific plant species, providing the ideal balance of vitamins and nutrients for enhanced cell growth and regeneration. This 10-liter formulation is commonly used in plant biotechnology, research, and tissue culture studies where large volumes of media are required to support extensive plant cell cultivation and genetic research experiments.
Catalog No. 50-006-59866 Supplier Alkali Scientific Supplier No. KOO002CP
Murashige & Skoog Medium with Vitamins, Van der Salm Modification, is a modified growth medium designed for large-scale plant tissue culture applications. With 44.6 grams of powder, it makes 10 liters of solution, supporting the growth and development of plant cells. The Van der Salm modification improves the medium for specific plant species, providing the ideal balance of vitamins and nutrients for enhanced cell growth and regeneration. This 10-liter formulation is commonly used in plant biotechnology, research, and tissue culture studies where large volumes of media are required to support extensive plant cell cultivation and genetic research experiments.