Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Murashige and Skoog Medium, Without Sucrose, Agar, IAA, and Kinetin

Catalog No. MP092610022 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 x 10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP092610022 1 x 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP092610022 Supplier MP Biomedicals™ Supplier No. 092610022
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Murashige and Skoog Medium, Without Sucrose, Agar, IAA, and Kinetin, Powder

  • Murashige and Skoog Medium is widely used plant tissue culture growth medium.
  • It has proven effective in the culture of tissue derived from moncotyledons and dicotyledons.
  • This medium, originally formulated to support tobacco callus, has been used to support callus initiation and growth, the growth of cells in suspension culture and the regeneration of shoots and plantlets from explants.
  • More recently, it has been used for micropropagation of ornamental, vegetable and fruit plants in research and commercial enterprises.
  • Murashige and Skoog Medium contains Murashige and Skoog salts, as well as a mixture of vitamins.
  • This modification of the original formula does not contain agar, sucrose, IAA, and Kinetin.
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Cell Culture Medium
For Use With (Application) Plant tissue culturegrowth
Quantity 1 x 10 L
Endotoxin Level <0.5 EU/mL
Formulation Component: Amino Acids-Glycine: 2.00000mg/L; Vitamins- myo-Inositol: 100.00000mg/L; Niacin: 0.50000mg/L; Pyridoxine HCl: 0.50000mg/L; Thiamine HCl 0.10000mg/L; Inorganic Salts-Ammonium Nitrate [NH4NO3]: 1650.00000mg/L; Boric Acid [H3BO3]: 6.20000mg/L; Calcium Chloride [CaCl2 2H2O] (anhyd): 332.0000mg/L; Cobalt Chloride [CoCl2 6H2O] Hexahydrate: 0.02500mg/L; Cupric Sulfate [CuSO4]: 0.01600mg/L; EDTA Fe Na: 36.70000mg/L; Magnesium Sulfate [MgSO4]: 180.70000mg/L; Manganese Sulfate [MnSO4 H2O] Monohydrate: 16.90000mg/L; Potassium Iodide [KI]: 0.83000mg/L; Potassium Nitrate [KNO3]: 1900.00000mg/L; Potassium Phosphate Monobasic [KH2PO4]: 170.00000mg/L; Sodium Molybdate [Na2MoO4 2H2O] Dihydrate: 0.25000mg/L; Zinc Sulfate [ZnSO4 7H2O] Heptahydrate: 8.60000mg/L
pH 4.80 to 5.65 (before additions)
Content And Storage Store at +4°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.