Learn More
Crescent Chemical Co Inc MURASHIGE & SKOOG MINIMAL ORGA
SDP
Catalog No. 50249136
Encompass_Preferred
Murashige and Skoog minimal organic powder medium 4,62 g/L Murashige and Skoog Plant Salts with i-inositol and thiamine hydrochloride. Without agar and without sucrose. Size - 5x10L Storage Conditions - +2 °C TO +8 °C Catalog Number - 47515.03
Catalog No. 50-249-136 Supplier Crescent Chemical Co Inc Supplier No. SE47515.03
Murashige and Skoog minimal organic powder medium 4,62 g/L Murashige and Skoog Plant Salts with i-inositol and thiamine hydrochloride. Without agar and without sucrose. Size - 5x10L Storage Conditions - +2 °C TO +8 °C Catalog Number - 47515.03