Learn More
Bioworld Murashige and Skoog(MS) Basal Salt (20X), 500 mL
Catalog No. 50254738
Encompass_Preferred
Murashige & Skoog (MS) Basal Salt ConcentrateContains Macro & Micronutrients as described by Murashige & Skoog (1962) In 20X Liquid ConcentratePlant tissue culture testedWorking Concentration: 50 mL/ LpH: 2.75–3.75
Catalog No. 50-254-738 Supplier Bioworld Supplier No. 306300871
Murashige & Skoog (MS) Basal Salt ConcentrateContains Macro & Micronutrients as described by Murashige & Skoog (1962) In 20X Liquid ConcentratePlant tissue culture testedWorking Concentration: 50 mL/ LpH: 2.75–3.75