Learn More
Bioworld Murashige and Skoog (MS) Medium, Macronutrients Only, 25L
Catalog No. 50254695
Encompass_Preferred
Contains only the macronutrients as described by Murashige & Skoog (1962). Micronutrients, Cytokines, Vitamins, Auxins or sucrose should be added as needed. Working Concentration (in g/L) 4.23, Powder. Shelf Life 4 years.
Catalog No. 50-254-695 Supplier Bioworld Supplier No. 306300702
Contains only the macronutrients as described by Murashige & Skoog (1962). Micronutrients, Cytokines, Vitamins, Auxins or sucrose should be added as needed. Working Concentration (in g/L) 4.23, Powder. Shelf Life 4 years.