Learn More
Bioworld Murashige and Skoog (MS) Medium, Macronutrients Only, 50L
Catalog No. 50254696
Encompass_Preferred
Contains only the macronutrients as described by Murashige & Skoog (1962). Micronutrients, Cytokines, Vitamins, Auxins or sucrose should be added as needed.
Catalog No. 50-254-696 Supplier Bioworld Supplier No. 306300703
Contains only the macronutrients as described by Murashige & Skoog (1962). Micronutrients, Cytokines, Vitamins, Auxins or sucrose should be added as needed.