Learn More
Bioworld Murashige and Skoog (MS) Medium Plates w/ Kanamycin, 50 Plate(s)
Catalog No. 50254677
Encompass_Preferred
Murashige & Skoog (MS) Medium PlatesPlates containing the Micronutrients and Macronutrients as describe by Murashige & Skoog (1967) with the addition of Kanamycin at 50 ug/mL
Catalog No. 50-254-677 Supplier Bioworld Supplier No. 306300622
Murashige & Skoog (MS) Medium PlatesPlates containing the Micronutrients and Macronutrients as describe by Murashige & Skoog (1967) with the addition of Kanamycin at 50 ug/mL