Learn More
Bioworld Murashige and Skoog(MS) Medium, w/ Sucrose and Agar, 25 L
Catalog No. 50254832
Encompass_Preferred
Murashige & Skoog (MS) Basal MediumContains Macroelements, Microelements, and Vitamins as described by Murashige & Skoog (1962) with the addition of Sucrose and Agar Add Cytokines and Auxins as needed Working Concentration: 42.4 g/L
Catalog No. 50-254-832 Supplier Bioworld Supplier No. 306301363
Murashige & Skoog (MS) Basal MediumContains Macroelements, Microelements, and Vitamins as described by Murashige & Skoog (1962) with the addition of Sucrose and Agar Add Cytokines and Auxins as needed Working Concentration: 42.4 g/L