Learn More
Bioworld Murashige and Skoog (MS) Medium w/ Gamborg B5 Vitamins, 25 L
Catalog No. 50254672
Encompass_Preferred
Murashige & Skoog (MS) MediumContains the Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Gamborg, et al. (1968)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant culture tissue testedWorking Concentration: 4.44 g/LpH: 3.5–4.5Soluble in water
Catalog No. 50-254-672 Supplier Bioworld Supplier No. 306300592
Murashige & Skoog (MS) MediumContains the Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Gamborg, et al. (1968)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant culture tissue testedWorking Concentration: 4.44 g/LpH: 3.5–4.5Soluble in water