Learn More
Bioworld Murashige and Skoog(MS) Medium w/ Van der Salm Modification w/ Vitamins, 25 L
Catalog No. 50254730
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Macronutrients, Micronutrients as described by Murashige & Skoog (1962) FeNaEDTA replaced by FeEDDHA as described by Van der Salm et al. (1994)Sucrose, Cytokines, and Auxins should be added as needed Working concentration: 4.36 g/LProvided as 1 unit to make 10L, 25L, or 50L of medium.Plant tissue culture tested
Catalog No. 50-254-730 Supplier Bioworld Supplier No. 306300823
Murashige & Skoog (MS) MediumContains Macronutrients, Micronutrients as described by Murashige & Skoog (1962) FeNaEDTA replaced by FeEDDHA as described by Van der Salm et al. (1994)Sucrose, Cytokines, and Auxins should be added as needed Working concentration: 4.36 g/LProvided as 1 unit to make 10L, 25L, or 50L of medium.Plant tissue culture tested