Learn More
Bioworld Murashige and Skoog (MS) Medium, w/ Gamborg B5 Vitamins 50ml
Catalog No. 50254673
Encompass_Preferred
Contains the Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Gamborg, et al. (1968). Sucrose, Cytokines, and Auxins should be added as needed. Plant culture tissue tested. Working Concentration: 4.44 g/L, pH: 3.5–4.5. Soluble in water 50l
Catalog No. 50-254-673 Supplier Bioworld Supplier No. 306300593
Contains the Macronutrients and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Gamborg, et al. (1968). Sucrose, Cytokines, and Auxins should be added as needed. Plant culture tissue tested. Working Concentration: 4.44 g/L, pH: 3.5–4.5. Soluble in water 50l