Learn More
Bioworld Murashige and Skoog (MS) Medium w/ Nitsch Vitamins, 50 L
Catalog No. 50254686
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1967) with addition of Vitamins as described by Nitsch & Nitsch (1969)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant tissue culture tested
Catalog No. 50-254-686 Supplier Bioworld Supplier No. 306300664
Murashige & Skoog (MS) MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1967) with addition of Vitamins as described by Nitsch & Nitsch (1969)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant tissue culture tested