Learn More
Bioworld Murashige and Skoog(MS) Medium w/ Finer & Nagasawa Modification, 25 L
Catalog No. 50254725
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Micronutrients and Macronutrients as described by Murashige & Skoog (1967) with 0.5 x concentration of NH4NO3Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.61 g/LSoluble in waterand 1.6 x concentrantion of KNO
Catalog No. 50-254-725 Supplier Bioworld Supplier No. 306300802
Add to Cart
Add to Cart
Murashige & Skoog (MS) MediumContains Micronutrients and Macronutrients as described by Murashige & Skoog (1967) with 0.5 x concentration of NH4NO3Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.61 g/LSoluble in waterand 1.6 x concentrantion of KNO