Learn More
Bioworld Murashige and Skoog (MS) Medium w/ Modified Vitamins, 50 L
Catalog No. 50254683
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Macronutrients, Micronutrients, and Modified Vitamins as described by Murashige & Skoog (1967)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant tissue culture tested
Catalog No. 50-254-683 Supplier Bioworld Supplier No. 306300654
Murashige & Skoog (MS) MediumContains Macronutrients, Micronutrients, and Modified Vitamins as described by Murashige & Skoog (1967)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant tissue culture tested