Learn More
Bioworld Murashige and Skoog(MS) Medium, Modification No. 5, 25 L
Catalog No. 50254720
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Micronutrients and Macronutrients with NH4NO3 replaced by NaNO3 as described by Murashige & Skoog (1967) Sucrose, Cytokinins, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-720 Supplier Bioworld Supplier No. 306300783
Murashige & Skoog (MS) MediumContains Micronutrients and Macronutrients with NH4NO3 replaced by NaNO3 as described by Murashige & Skoog (1967) Sucrose, Cytokinins, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested