Learn More
Bioworld Murashige and Skoog(MS) Medium, BC Potato, 50 L
Catalog No. 50254843
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962)Contains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTASucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.30 g/LpH: 4.25–5.25Soluble in water
Catalog No. 50-254-843 Supplier Bioworld Supplier No. 306301424
Murashige & Skoog (MS) MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962)Contains Ferric Sodium EDTA in place of Ferrous Sulfate and Disodium EDTASucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.30 g/LpH: 4.25–5.25Soluble in water