Learn More
Bioworld Murashige and Skoog (MS) Medium w/ Vitamins, Buffered, 25 L
Catalog No. 50254691
Encompass_Preferred
Murashige & Skoog (MS) MediumContains the Micronutrients, Macronutrients, and Vitamins as described by Murashige & Skoog (1962)Buffered with MESSucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Conentration: 4.9 g/L pH: 5.5−6.7 Soluble in water
Catalog No. 50-254-691 Supplier Bioworld Supplier No. 306300683
Murashige & Skoog (MS) MediumContains the Micronutrients, Macronutrients, and Vitamins as described by Murashige & Skoog (1962)Buffered with MESSucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Conentration: 4.9 g/L pH: 5.5−6.7 Soluble in water