Learn More
Bioworld Murashige and Skoog(MS) Medium, Modification No. 2, 25 L
Catalog No. 50254704
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Micronutrients and 3/4 Macronutrients as described by Murashige & Skoog (1967) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of medium Plant tissue culture tested
Catalog No. 50-254-704 Supplier Bioworld Supplier No. 306300733
Murashige & Skoog (MS) MediumContains Micronutrients and 3/4 Macronutrients as described by Murashige & Skoog (1967) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of medium Plant tissue culture tested