Learn More
Bioworld Murashige and Skoog(MS) Medium, Modification No. 4, 25 L
Catalog No. 50254716
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Macronutrients and Micronutrients with No NH4NO3 as described by Murashige & Skoog (1962)Sucrose, Cytokines, Vitamins, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking concentration 2.7 g/L
Catalog No. 50-254-716 Supplier Bioworld Supplier No. 306300773
Murashige & Skoog (MS) MediumContains Macronutrients and Micronutrients with No NH4NO3 as described by Murashige & Skoog (1962)Sucrose, Cytokines, Vitamins, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking concentration 2.7 g/L