Learn More
Bioworld Murashige and Skoog(MS) Medium, Modification No. 1 w/ Vitamins, 25 L
Catalog No. 50254701
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Micronutrients, 1/2 Macronutrients, and Vitamins as described by Murashige & Skoog (1967)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-701 Supplier Bioworld Supplier No. 306300723
Murashige & Skoog (MS) MediumContains Micronutrients, 1/2 Macronutrients, and Vitamins as described by Murashige & Skoog (1967)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested