Learn More
Bioworld Murashige and Skoog(MS) Medium w/ Van der Salm Modification, 50 L
Catalog No. 50254728
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Macronutrients, Micronutrients and vitamins as described by Murashige & Skoog (1962) with FeNaEDTA replaced by FeEDDHA (a highly stabile chelate providing a source of iron easily absorbed by plants) as described by Van der Salm et al. (1994)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium.Plant tissue culture testedWorking Concentration: 4.46 g/LpH: ≈ 3.5-4.5Soluble in water
Catalog No. 50-254-728 Supplier Bioworld Supplier No. 306300813
Murashige & Skoog (MS) MediumContains Macronutrients, Micronutrients and vitamins as described by Murashige & Skoog (1962) with FeNaEDTA replaced by FeEDDHA (a highly stabile chelate providing a source of iron easily absorbed by plants) as described by Van der Salm et al. (1994)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium.Plant tissue culture testedWorking Concentration: 4.46 g/LpH: ≈ 3.5-4.5Soluble in water