Learn More
Bioworld Murashige and Skoog (MS) Medium, 100 L
Catalog No. 50254670
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Micronutrients and Macronutrients as described by Murashige & Skoog (1962) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of medium; or as 1 kg of powderPlant tissue culture testedWorking Concentration: 4.3 g/L
Catalog No. 50-254-670 Supplier Bioworld Supplier No. 306300585
Murashige & Skoog (MS) MediumContains Micronutrients and Macronutrients as described by Murashige & Skoog (1962) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of medium; or as 1 kg of powderPlant tissue culture testedWorking Concentration: 4.3 g/L