Learn More
Alkali Scientific Murashige & Skoog MS Medium, 21.5 Grams of Powder, Makes 5 Liters of Solution
SDP
Catalog No. 5000661660
Encompass_Preferred
Murashige & Skoog (MS) Medium, designed in powder form, contains 21.5 grams of the medium powder to make 5 liters of solution. This medium is an essential tool for plant tissue culture, supporting the growth of a wide range of plants, including herbs, fruits, and ornamental species. The MS formulation includes essential macro and micronutrients, plant growth regulators, and vitamins necessary for plant regeneration, root formation, and shoot proliferation. It is frequently used for plant breeding, propagation, and in vitro conservation studies. This product is especially useful for researchers and agricultural professionals working with small- to medium-scale plant tissue culture projects.
Catalog No. 50-006-61660 Supplier Alkali Scientific Supplier No. MUR005CP
Murashige & Skoog (MS) Medium, designed in powder form, contains 21.5 grams of the medium powder to make 5 liters of solution. This medium is an essential tool for plant tissue culture, supporting the growth of a wide range of plants, including herbs, fruits, and ornamental species. The MS formulation includes essential macro and micronutrients, plant growth regulators, and vitamins necessary for plant regeneration, root formation, and shoot proliferation. It is frequently used for plant breeding, propagation, and in vitro conservation studies. This product is especially useful for researchers and agricultural professionals working with small- to medium-scale plant tissue culture projects.