Learn More
Bioworld Murashige and Skoog (MS) Medium 25L
Catalog No. 50254668
Encompass_Preferred
Sucrose, Cytokines, Vitamins, and Auxins should be added as needed. Murashige & Skoog (MS) Medium is plant tissue culture tested with a working concentration of 4.33 g/L. Murashige & Skoog (MS) Basal Salt Medium is provided as 1 unit to make 25L or 50L of medium.
Catalog No. 50-254-668 Supplier Bioworld Supplier No. 306300583
Sucrose, Cytokines, Vitamins, and Auxins should be added as needed. Murashige & Skoog (MS) Medium is plant tissue culture tested with a working concentration of 4.33 g/L. Murashige & Skoog (MS) Basal Salt Medium is provided as 1 unit to make 25L or 50L of medium.