Learn More
Bioworld Murashige and Skoog(MS) Modified Medium w/o Potassium Phosphate, 25 L
Catalog No. 50255002
Encompass_Preferred
Contains Micronutrients and Macronutrients as described by Murashige & Skoog (1962) modified addition of Sodium phosphate monobasic instead of Potassium phosphate monobasic, FeNaEDTA instead of Ferrous sulfate and NaEDTA.Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 5.69 g/L
Catalog No. 50-255-002 Supplier Bioworld Supplier No. 306302012
Contains Micronutrients and Macronutrients as described by Murashige & Skoog (1962) modified addition of Sodium phosphate monobasic instead of Potassium phosphate monobasic, FeNaEDTA instead of Ferrous sulfate and NaEDTA.Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 5.69 g/L