Learn More
Bioworld Murashige and Skoog(MS) Modified Medium w/o Nitrogen, 50 L
Catalog No. 50255000
Encompass_Preferred
Murashige and Skoog Medium w/o NitrogenContains Micronutrients and Macronutrients as described by Murashige & Skoog (1962) but without Ammonium nitrate and Potassium nitrate.Sucrose, Cytokines, Vitamins, and Auxins should be added as needed. Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 0.78 g/L
Catalog No. 50-255-000 Supplier Bioworld Supplier No. 306302003
Murashige and Skoog Medium w/o NitrogenContains Micronutrients and Macronutrients as described by Murashige & Skoog (1962) but without Ammonium nitrate and Potassium nitrate.Sucrose, Cytokines, Vitamins, and Auxins should be added as needed. Provided as 1 unit to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 0.78 g/L