Learn More
Bioworld Murashige and Skoog(MS) Modified Medium, Arabidopsis Culture Medium, 50 L
Catalog No. 50254845
Encompass_Preferred
Murashige & Skoog (MS) Modified MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962)Contains additional nutrients specific for Arabidopsis culturingProvided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 24.44 g/LSoluble in water
Catalog No. 50-254-845 Supplier Bioworld Supplier No. 306301433
Murashige & Skoog (MS) Modified MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962)Contains additional nutrients specific for Arabidopsis culturingProvided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 24.44 g/LSoluble in water