Learn More
Bioworld Murashige and Skoog(MS) Modified Medium w/ ½Macro & ½Micro, 50 L
Catalog No. 50255008
Encompass_Preferred
Murashige & Skoog (MS) Modified Medium w/ ½ Macro and ½MicronutrientsContains ½ Macronutrients and ½ Micronutrients as described by Murashige & Skoog (1962)Provided as 1 unit to make 25L or 50L of mediumPlant tissue culture testedWorking Concentration: 2.17 g/LSoluble in water
Catalog No. 50-255-008 Supplier Bioworld Supplier No. 306302032
Murashige & Skoog (MS) Modified Medium w/ ½ Macro and ½MicronutrientsContains ½ Macronutrients and ½ Micronutrients as described by Murashige & Skoog (1962)Provided as 1 unit to make 25L or 50L of mediumPlant tissue culture testedWorking Concentration: 2.17 g/LSoluble in water