Learn More
Bioworld Murashige and Skoog(MS) Modified Medium w/ NAA & BA, 25 L
Catalog No. 50255019
Encompass_Preferred
Contains the Macronutrients, Micronutrients, and Vitamins as described by Murashige & Skoog (1962) modified with 0.1mg/L of 6-BAP and α-NAA and 30g/L Sucrose.Sold in individual containers with enough medium to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 34.44 g/LSoluble in water
Catalog No. 50-255-019 Supplier Bioworld Supplier No. 306302072
Add to Cart
Add to Cart
Contains the Macronutrients, Micronutrients, and Vitamins as described by Murashige & Skoog (1962) modified with 0.1mg/L of 6-BAP and α-NAA and 30g/L Sucrose.Sold in individual containers with enough medium to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 34.44 g/LSoluble in water