Learn More
Bioworld Murashige and Skoog (MS) Modified Medium, w/ IAA, 25 L
Catalog No. 50254850
Encompass_Preferred
Murashige & Skoog (MS) Modified MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) modified with addition of Sodium phosphate monobasic, Sucrose, 2iP, Adenine Hemisulfate, and IAAProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 34.69 g/LSoluble in water
Catalog No. 50-254-850 Supplier Bioworld Supplier No. 306301453
Murashige & Skoog (MS) Modified MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) modified with addition of Sodium phosphate monobasic, Sucrose, 2iP, Adenine Hemisulfate, and IAAProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 34.69 g/LSoluble in water