Learn More
Bioworld Murashige and Skoog(MS) Modified Medium w/o Nitrogen, Potassium or Phosphates, 25 L
Catalog No. 50255013
Encompass_Preferred
Contains the Macronutrients, Micronutrients as described by Murashige & Skoog (1962) but without Ammonium nitrate, Potassium nitrate or Potassium phosphate monbasicCytokines, Sucrose, and Auxins can be added as needed Sold in individual containers with enough medium to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 0.61 g/LpH: 3.8−4.8Soluble in water
Catalog No. 50-255-013 Supplier Bioworld Supplier No. 306302052
Contains the Macronutrients, Micronutrients as described by Murashige & Skoog (1962) but without Ammonium nitrate, Potassium nitrate or Potassium phosphate monbasicCytokines, Sucrose, and Auxins can be added as needed Sold in individual containers with enough medium to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 0.61 g/LpH: 3.8−4.8Soluble in water