Learn More
Bioworld Murashige and Skoog(MS) Modified Medium, w/ Adenine Hemisulfate, 50 L
Catalog No. 50254848
Encompass_Preferred
Murashige & Skoog (MS) Modified MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962), vitamins as described by linsmaer and Skoog (1965), modified with addition of Adenine Hemisulfate (80mg/L)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.51 g/LSoluble in water Synonym: Murashige Begonia Multiplication Medium
Catalog No. 50-254-848 Supplier Bioworld Supplier No. 306301444
Murashige & Skoog (MS) Modified MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1962), vitamins as described by linsmaer and Skoog (1965), modified with addition of Adenine Hemisulfate (80mg/L)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.51 g/LSoluble in water Synonym: Murashige Begonia Multiplication Medium