Learn More
Bioworld Murashige and Skoog(MS) Modified Vitamin Powder, 100 mL
Catalog No. 50254736
Encompass_Preferred
Murashige & Skoog (MS) Modified Vitamin PowderContains the Vitamins as described by Murashige & Skoog (1962) modified with additional ThiaminePlant tissue culture testedWorking Concentration: 10.40 g/ 100 mLpH: 3.5–4.5Soluble in water
Catalog No. 50-254-736 Supplier Bioworld Supplier No. 306300861
Murashige & Skoog (MS) Modified Vitamin PowderContains the Vitamins as described by Murashige & Skoog (1962) modified with additional ThiaminePlant tissue culture testedWorking Concentration: 10.40 g/ 100 mLpH: 3.5–4.5Soluble in water