Learn More
Bioworld Murashige and Skoog(MS) Modified Multiplication Medium C, 50 L
Catalog No. 50254854
Encompass_Preferred
Murashige & Skoog (MS) Modified Multiplication Medium CContains Macro and Micronutrients as described by Murashige & Skoog (1962) and the Vitamins as described by Linsmaier and Skoog (1965) modified with addition of Kinetin, Adenine Hemisulfate, Sodium Phosphate Monobasic, NAA, and SucroseCytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 34.66 g/LSoluble in water
Catalog No. 50-254-854 Supplier Bioworld Supplier No. 306301464
Murashige & Skoog (MS) Modified Multiplication Medium CContains Macro and Micronutrients as described by Murashige & Skoog (1962) and the Vitamins as described by Linsmaier and Skoog (1965) modified with addition of Kinetin, Adenine Hemisulfate, Sodium Phosphate Monobasic, NAA, and SucroseCytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 34.66 g/LSoluble in water