Learn More
Bioworld Murashige and Skoog(MS) Modified Medium, 25 L
Catalog No. 50255011
Encompass_Preferred
Murashige & Skoog (MS) Modified Medium with ½ NH4N03, ½ KNO3 and ½CaCl2Contains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) but with half concentration of Ammonium nitrate, potassium nitrate and Calcium nitrateProvided as 1 unit to make 25L or 50L of mediumPlant tissue culture testedWorking Concentration: 2.39 g/LSoluble in water
Catalog No. 50-255-011 Supplier Bioworld Supplier No. 306302042
Murashige & Skoog (MS) Modified Medium with ½ NH4N03, ½ KNO3 and ½CaCl2Contains Macronutrients and Micronutrients as described by Murashige & Skoog (1962) but with half concentration of Ammonium nitrate, potassium nitrate and Calcium nitrateProvided as 1 unit to make 25L or 50L of mediumPlant tissue culture testedWorking Concentration: 2.39 g/LSoluble in water